1-Chloro-2,3-difluoro-5-nitrobenzene 98%

Supplier: Ambeed
Warning

A290221-1G A290221-5G
77289-780EA 18 USD
77289-780 77289-782
1-Chloro-2,3-difluoro-5-nitrobenzene 98%
1-Chloro-2,3-difluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClF₂NO₂
MDL Number: MFCD06658261
CAS Number: 53780-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR