2,2-Bis[(4S)-4-benzyl-2-oxazolin-2-yl]propane ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A290143-100MG
A290143-1G
A290143-250MG
77250-654EA
24.72
USD
77250-654
77250-656
77250-658
2,2-Bis[(4S)-4-benzyl-2-oxazolin-2-yl]propane ≥98%, ee 99%
2,2-Bis[(4S)-4-benzyl-2-oxazolin-2-yl]propane
Precaución: for Research Use Only
Formula:
C₂₃H₂₆N₂O₂ MW: 362.46 g/mol |
MDL Number:
MFCD30343613 CAS Number: 176706-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...