2-Methyl-3-nitrobenzyl alcohol 97%

Supplier: Ambeed
Warning

A289928-100G A289928-10G A289928-1G A289928-250MG A289928-25G
77250-644EA 672.59 USD
77250-644 77250-646 77250-648 77250-650 77250-652
2-Methyl-3-nitrobenzyl alcohol 97%
2-Methyl-3-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.162 g/mol
MDL Number: MFCD00007166
CAS Number: 23876-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR