3-(Trifluoromethyl)pyridine-4-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Trifluoromethyl)isonicotinamide

A289688-100MG A289688-1G A289688-250MG
77239-032EA 36.05 USD
77239-032 77239-034 77239-036
3-(Trifluoromethyl)pyridine-4-carboxamide 97%
3-(Trifluoromethyl)pyridine-4-carboxamide

Precaución: for Research Use Only

Formula: C₇H₅F₃N₂O
MW: 190.123 g/mol
MDL Number: MFCD22684694
CAS Number: 1416713-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR