(S)-Hexahydropyrazino[2,1-c][1,4]oxazin-4(3H)-one hydrochloride 96%

Supplier: Ambeed
Warning

A289581-100MG A289581-250MG
77262-814EA 332.69 USD
77262-814 77262-816
(S)-Hexahydropyrazino[2,1-c][1,4]oxazin-4(3H)-one hydrochloride 96%
(S)-Hexahydropyrazino[2,1-c][1,4]oxazin-4(3H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃ClN₂O₂
MW: 192.64 g/mol
MDL Number: MFCD30470764
CAS Number: 1383427-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR