4-(4-Methoxy-2,3,6-trimethylphenyl)but-3-en-2-one 97%
Supplier: Ambeed
|
Warning
|
A289510-10G
A289510-1G
A289510-250MG
A289510-5G
77289-772EA
80.34
USD
77289-772
77289-774
77289-776
77289-778
4-(4-Methoxy-2,3,6-trimethylphenyl)but-3-en-2-one 97%
4-(4-Methoxy-2,3,6-trimethylphenyl)but-3-en-2-one
Precaución: for Research Use Only
Formula:
C₁₄H₁₈O₂ MW: 218.296 g/mol |
MDL Number:
MFCD00449197 CAS Number: 54757-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...