2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

A289440-1G A289440-250MG
77262-810EA 61 USD
77262-810 77262-812
2-Fluoro-5-(trifluoromethyl)benzenesulfonyl chloride 98%
2-Fluoro-5-(trifluoromethyl)benzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃ClF₄O₂S
MDL Number: MFCD01090999
CAS Number: 1744-43-0
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR