4-Hydroxyisophthalaldehyde 98%

Supplier: Ambeed
Warning

A289333-1G A289333-250MG A289333-25G A289333-5G
77239-012EA 54 USD
77239-012 77239-014 77239-016 77239-018
4-Hydroxyisophthalaldehyde 98%
4-Hydroxyisophthalaldehyde

Precaución: for Research Use Only

Formula: C₈H₆O₃
MDL Number: MFCD00003334
CAS Number: 3328-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR