2,5-Dichloroanisole 98%

Supplier: Ambeed
Warning

A289135-100G A289135-10G A289135-1G A289135-25G
77250-620EA 94 USD
77250-620 77250-622 77250-624 77250-626
2,5-Dichloroanisole 98%
2,5-Dichloroanisole

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O
MDL Number: MFCD00061123
CAS Number: 1984-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR