3,3'-Dihydroxybenzidine 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol

A289113-10G A289113-1G A289113-25G A289113-5G A289113-100G
77277-152EA 67.98 USD
77277-152 77277-154 77277-156 77277-158 77568-588
3,3'-Dihydroxybenzidine 97%
3,3'-Dihydroxybenzidine

Precaución: For research use only.

Formula: C₁₂H₁₂N₂O₂
MW: 216.24 g/mol
MDL Number: MFCD00039149
CAS Number: 2373-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR