(2-Chloro-5-fluorophenyl)hydrazine hydrochloride 98%

Supplier: Ambeed
Warning

A289075-250MG A289075-5G
77262-798EA 5.15 USD
77262-798 77262-800
(2-Chloro-5-fluorophenyl)hydrazine hydrochloride 98%
(2-Chloro-5-fluorophenyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₇Cl₂FN₂
MW: 197.0400 g/mol
MDL Number: MFCD03094167
CAS Number: 502496-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR