3-Phosphonopropionic acid 98%

Supplier: Ambeed
Danger

Synonyms: 3-Phosphonopropanoic acid

A289063-1G A289063-25G A289063-5G
77289-766EA 14 USD
77289-766 77289-768 77289-770
3-Phosphonopropionic acid 98%
3-Phosphonopropionic acid

Precaución: for Research Use Only

Formula: C₃H₇O₅P
MW: 154.06 g/mol
MDL Number: MFCD00002770
CAS Number: 5962-42-5
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR