(9H-Fluoren-9-yl)methyl hydroxycarbamate 97%

Supplier: Ambeed
Warning

A288749-1G A288749-250MG
77262-780EA 6 USD
77262-780 77262-782
(9H-Fluoren-9-yl)methyl hydroxycarbamate 97%
(9H-Fluoren-9-yl)methyl hydroxycarbamate

Precaución: for Research Use Only

Formula: C₁₅H₁₃NO₃
MDL Number: MFCD01862911
CAS Number: 190656-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR