5-Chloroindole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-1H-indole-2-carboxylic acid

A288544-100G A288544-10G A288544-1G A288544-25G A288544-500G A288544-5G
77277-140EA 331.66 USD
77277-140 77277-142 77277-144 77277-146 77277-148 77277-150
5-Chloroindole-2-carboxylic acid 97%
5-Chloroindole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MW: 195.6 g/mol
MDL Number: MFCD00005613
CAS Number: 10517-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR