Sodium bis(trifluoromethylsulfonyl)imide 98%

Supplier: Ambeed
Danger

Synonyms: Bis(trifluoromethanesulphonyl)imide sodium salt, Sodium triflimide, Sodium trifluoromethanesulphonimide, Sodium Bis(trifluoromethanesulphonyl)imide

A288402-1G A288402-25G A288402-5G
77262-778EA 6 USD
77262-778 77550-762 77550-764
Sodium bis(trifluoromethylsulfonyl)imide 98%
Sodium bis(trifluoromethylsulfonyl)imide

Precaución: for Research Use Only

Formula: C₂F₆NNaO₄S₂
MDL Number: MFCD14156021
CAS Number: 91742-21-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR