1-(Cyclopropylmethyl)-3-hydroxyquinazoline-2,4(1H,3H)-dione 98%

Supplier: Ambeed
Warning

A288260-1MG A288260-25MG A288260-50MG A288260-10MG A288260-5MG
77262-772EA 15 USD
77262-772 77262-774 77262-776 77748-560 77748-561
1-(Cyclopropylmethyl)-3-hydroxyquinazoline-2,4(1H,3H)-dione 98%
1-(Cyclopropylmethyl)-3-hydroxyquinazoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₃
MDL Number: MFCD31812378
CAS Number: 1995893-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR