1-Ethynyl-4-propoxybenzene 97%

Supplier: Ambeed
Warning

A287990-10G A287990-1G A287990-5G
77239-002EA 94.99 USD
77239-002 77239-004 77239-006
1-Ethynyl-4-propoxybenzene 97%
1-Ethynyl-4-propoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₂O
MDL Number: MFCD00173879
CAS Number: 39604-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR