2-Bromo-1,1-diethoxypropane 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromopriopionaldehydediethylacetal

A287729-10G A287729-1G A287729-25G A287729-5G
77262-758EA 131.84 USD
77262-758 77262-760 77262-762 77262-764
2-Bromo-1,1-diethoxypropane 98%
2-Bromo-1,1-diethoxypropane

Precaución: for Research Use Only

Formula: (H₃CCH₂O)₂HCBrCHCH₃
MW: 211.1 g/mol
MDL Number: MFCD00152299
CAS Number: 3400-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR