Adamantan-2-amine 95%

Supplier: Ambeed
Warning

A287681-10G A287681-1G A287681-250MG A287681-5G
77262-750EA 105 USD
77262-750 77262-752 77262-754 77262-756
Adamantan-2-amine 95%
Adamantan-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₇N
MDL Number: MFCD00078225
CAS Number: 13074-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR