Sulfatinib 99+%

Supplier: Ambeed
Warning

Synonyms: N-(2-(Dimethylamino)ethyl)-1-(3-((4-((2-methyl-1H-indol-5-yl)oxy)pyrimidin-2-yl)amino)phenyl)methanesulphonamide

A287603-100MG A287603-10MG A287603-5MG
77250-606EA 32.96 USD
77250-606 77250-608 77250-610
Sulfatinib 99+%
Sulfatinib

Precaución: for Research Use Only

Formula: C₂₄H₂₈N₆O₃S
MW: 480.59 g/mol
MDL Number: MFCD28359080
CAS Number: 1308672-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR