2-((3-(4-Cyanonaphthalen-1-yl)pyridin-4-yl)thio)-2-methylpropanoic acid 98+%

Supplier: Ambeed
Warning

A287467-10MG A287467-1MG A287467-25MG A287467-50MG A287467-5MG
77238-988EA 41 USD
77238-988 77238-990 77238-992 77238-994 77238-996
2-((3-(4-Cyanonaphthalen-1-yl)pyridin-4-yl)thio)-2-methylpropanoic acid 98+%
2-((3-(4-Cyanonaphthalen-1-yl)pyridin-4-yl)thio)-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₆N₂O₂S
MDL Number: MFCD28963965
CAS Number: 1352792-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR