(R)-3-(6-((4-Chlorophenyl)sulfonamido)-2-methyl-5,6,7,8-tetrahydronaphthalen-1-yl)propanoic acid 98%

Supplier: Ambeed
Warning

A287410-10MG A287410-1MG
77238-984EA 62.99 USD
77238-984 77238-986
(R)-3-(6-((4-Chlorophenyl)sulfonamido)-2-methyl-5,6,7,8-tetrahydronaphthalen-1-yl)propanoic acid 98%
(R)-3-(6-((4-Chlorophenyl)sulfonamido)-2-methyl-5,6,7,8-tetrahydronaphthalen-1-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₀H₂₂ClNO₄S
MDL Number: MFCD28411833
CAS Number: 165538-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR