4-Chloro-7-methylthieno[3,2-d]pyrimidine 97%

Supplier: Ambeed
Warning

A287226-100G A287226-1G A287226-250MG A287226-25G A287226-5G
77289-752EA 954.81 USD
77289-752 77289-754 77289-756 77289-758 77289-760
4-Chloro-7-methylthieno[3,2-d]pyrimidine 97%
4-Chloro-7-methylthieno[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂S
MDL Number: MFCD00205202
CAS Number: 175137-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR