Edicotinib 97%

Supplier: Ambeed
Warning

Synonyms: 5-Cyano-N-(2-(4,4-dimethylcyclohex-1-en-1-yl)-6-(2,2,6,6-tetramethyltetrahydro-2H-pyran-4-yl)pyridin-3-yl)-1H-imidazole-2-carboxamide, PRV 6527, JNJ 40346527, 4-cyano-N-[2-(4,4-dimethylcyclohex-1-en-1-yl)-6-(2,2,6,6-tetramethyloxan-4-yl)pyridin-3-yl]-1H-imidazole-2-carboxamide

A287103-10MG A287103-1MG A287103-5MG A287103-50MG
77250-596EA 91.67 USD
77250-596 77250-598 77250-600 77746-527
Edicotinib 97%
Edicotinib

Precaución: for Research Use Only

Formula: C₂₇H₃₅N₅O₂
MW: 461.61 g/mol
MDL Number: MFCD30489233
CAS Number: 1142363-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR