2-Methanesulfonylaminobenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Methylsulphonamido)benzoic acid

A286978-1G A286978-5G A286978-250MG
77262-746EA 47.38 USD
77262-746 77262-748 77748-555
2-Methanesulfonylaminobenzoic acid 98%
2-Methanesulfonylaminobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₄S
MW: 215.23 g/mol
MDL Number: MFCD00590092
CAS Number: 162787-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR