BPTU 98+%

Supplier: Ambeed
Warning

Synonyms: 1-(2-(2-(tert-Butyl)phenoxy)pyridin-3-yl)-3-(4-(trifluoromethoxy)phenyl)urea, N-[2-[2-(1,1-Dimethylethyl)phenoxy]-3-pyridinyl]-N'-[4-(trifluoromethoxy)phenyl]urea, BMS-646786

A286791-10MG A286791-1MG A286791-25MG A286791-50MG A286791-5MG
77277-084EA 28.84 USD
77277-084 77277-086 77277-088 77277-090 77277-092
BPTU 98+%
BPTU

Precaución: for Research Use Only

Formula: C₂₃H₂₂F₃N₃O₃
MW: 445.43 g/mol
MDL Number: MFCD28386231
CAS Number: 870544-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR