3-Methyl-1,3-thiazolane-2,4-dione 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methylthiazolidine-2,4-dione

A286790-1G A286790-250MG A286790-5G
77277-078EA 62.83 USD
77277-078 77277-080 77277-082
3-Methyl-1,3-thiazolane-2,4-dione 95%
3-Methyl-1,3-thiazolane-2,4-dione

Precaución: for Research Use Only

Formula: C₄H₅NO₂S
MW: 131.15 g/mol
MDL Number: MFCD02194457
CAS Number: 16312-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR