(3-Fluoro-4-hydroxyphenyl)acetic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Fluoro-4-hydroxyphenylacetic acid
A286776-5G
A286776-1G
77238-980EA
301.79
USD
77238-980
77746-524
(3-Fluoro-4-hydroxyphenyl)acetic acid 95%
(3-Fluoro-4-hydroxyphenyl)acetic acid
Precaución: for Research Use Only
Formula:
C₈H₇FO₃ MW: 170.14 g/mol |
MDL Number:
MFCD00004348 CAS Number: 458-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...