5-Fluoro-1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile 98%
Supplier: Ambeed
|
Warning
|
A286628-100MG
A286628-1G
A286628-250MG
A286628-5G
77262-732EA
15.45
USD
77262-732
77262-734
77262-736
77262-738
5-Fluoro-1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile 98%
5-Fluoro-1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₁₄H₈F₂N₄ MW: 270.23 g/mol |
MDL Number:
MFCD28502339 CAS Number: 1350653-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...