2-Bromo-6-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

A286615-10G A286615-25G A286615-5G A286615-1G
77277-072EA 22 USD
77277-072 77277-074 77277-076 77748-554
2-Bromo-6-fluorophenylboronic acid 97%
2-Bromo-6-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
MDL Number: MFCD05664309
CAS Number: 913835-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR