3-(tert-Butyl)-2-hydroxy-5-iodobenzaldehyde 97%

Supplier: Ambeed
Warning

A286524-250MG A286524-5G
77250-592EA 41.2 USD
77250-592 77250-594
3-(tert-Butyl)-2-hydroxy-5-iodobenzaldehyde 97%
3-(tert-Butyl)-2-hydroxy-5-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃IO₂
MW: 304.12 g/mol
MDL Number: MFCD27929482
CAS Number: 83816-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR