Potassium (2-benzyloxyethyl)trifluoroborate 95%

Supplier: Ambeed
Warning

Synonyms: Potassium (2-(benzyloxy)ethyl)trifluoroborate

A286497-100MG A286497-1G A286497-250MG A286497-5G
77238-974EA 13 USD
77238-974 77238-976 77238-978 77616-228
Potassium (2-benzyloxyethyl)trifluoroborate 95%
Potassium (2-benzyloxyethyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₉H₁₁BF₃O·K
MDL Number: MFCD22684708
CAS Number: 1408168-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR