4-Fluoro-2-methyl-6-nitroaniline 97%

Supplier: Ambeed
Warning

A285696-100G A285696-10G A285696-1G A285696-25G A285696-5G
77250-580EA 214.24 USD
77250-580 77250-582 77250-584 77250-586 77250-588
4-Fluoro-2-methyl-6-nitroaniline 97%
4-Fluoro-2-methyl-6-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇FN₂O₂
MDL Number: MFCD09152778
CAS Number: 147285-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR