3-(Naphthalen-2-yl)-1-phenyl-1H-pyrazole-4-carbaldehyde 97%

Supplier: Ambeed
Warning

A285589-100MG A285589-1G A285589-250MG
77262-710EA 5 USD
77262-710 77262-712 77262-714
3-(Naphthalen-2-yl)-1-phenyl-1H-pyrazole-4-carbaldehyde 97%
3-(Naphthalen-2-yl)-1-phenyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₂₀H₁₄N₂O
MDL Number: MFCD02918303
CAS Number: 36640-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR