(6-Chloro-1H-benzotriazol-1-yloxy)tripyrrolidinophosphonium Hexafluorophosphate 95%

Supplier: Ambeed
Warning

Synonyms: ((6-Chloro-1H-benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate, TPTDP

A285552-25G A285552-5G
77250-576EA 22 USD
77250-576 77250-578
(6-Chloro-1H-benzotriazol-1-yloxy)tripyrrolidinophosphonium Hexafluorophosphate 95%
(6-Chloro-1H-benzotriazol-1-yloxy)tripyrrolidinophosphonium Hexafluorophosphate

Precaución: for Research Use Only

Formula: C₁₈H₂₇ClF₆N₆OP₂
MW: 554.84 g/mol
MDL Number: MFCD14560589
CAS Number: 893413-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR