2-Amino-3-bromo-5-iodopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-iodopyridin-2-amine

A285530-1G A285530-250MG A285530-5G A285530-25G
77277-068EA 12 USD
77277-068 77277-070 77616-220 77748-553
2-Amino-3-bromo-5-iodopyridine 98%
2-Amino-3-bromo-5-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₄BrIN₂
MDL Number: MFCD08272065
CAS Number: 697300-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR