2-Fluoro-N-methoxy-N-methylbenzamide 95%

Supplier: Ambeed
Warning

A285446-1G A285446-5G
77262-706EA 37.08 USD
77262-706 77262-708
2-Fluoro-N-methoxy-N-methylbenzamide 95%
2-Fluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
MDL Number: MFCD02684304
CAS Number: 198967-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR