4-Bromo-2-chloro-3-methylpyridine 97%

Supplier: Ambeed
Warning

A285403-1G A285403-5G
77262-702EA 109.18 USD
77262-702 77262-704
4-Bromo-2-chloro-3-methylpyridine 97%
4-Bromo-2-chloro-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClN
MDL Number: MFCD09839276
CAS Number: 128071-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR