1-Propylpiperazine dihydrobromide 98%

Supplier: Ambeed
Warning

A285167-10G A285167-1G A285167-25G A285167-5G
77238-966EA 31.93 USD
77238-966 77238-968 77238-970 77238-972
1-Propylpiperazine dihydrobromide 98%
1-Propylpiperazine dihydrobromide

Precaución: for Research Use Only

Formula: C₇H₁₆N₂
MW: 290.04 g/mol
MDL Number: MFCD00051992
CAS Number: 64262-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR