5-Fluoro-o-toluic acid 98%

Supplier: Ambeed
Warning

A285165-10G A285165-1G A285165-500G A285165-100G
77277-054EA 13.39 USD
77277-054 77277-056 77277-058 77568-576
5-Fluoro-o-toluic acid 98%
5-Fluoro-o-toluic acid

Precaución: For research use only.

Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD00042294
CAS Number: 33184-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR