Methyl-1-boc-azetidine-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 1-Boc-azetidine-2-carboxylate, 1-Boc-Azetidine-2-carboxylic acid methyl ester
A284893-10G
A284893-1G
A284893-25G
A284893-5G
77238-954EA
188.49
USD
77238-954
77238-956
77238-958
77238-960
Methyl-1-boc-azetidine-2-carboxylate 97%
Methyl-1-boc-azetidine-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₄ MW: 215.2463 g/mol |
MDL Number:
MFCD06657099 CAS Number: 255882-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...