2,7-Diazaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2,7-diazaspiro[3.5]nonane-7-carboxylate

A284697-100MG A284697-250MG A284697-5G A284697-25G A284697-1G
77250-554EA 18.54 USD
77250-554 77250-556 77550-748 77571-770 77616-216
2,7-Diazaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 95%
2,7-Diazaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MW: 226.32 g/mol
MDL Number: MFCD09839640
CAS Number: 896464-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR