2-Methyl-3-heptanone 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Butyl Isopropyl ketone, 2-Methylheptan-3-one
A284658-1G
A284658-5G
77277-034EA
35.02
USD
77277-034
77277-036
2-Methyl-3-heptanone 98%
2-Methyl-3-heptanone
Precaución: for Research Use Only
Formula:
C₈H₁₆O MW: 128.21 g/mol |
MDL Number:
MFCD00009481 CAS Number: 13019-20-0 UN: 1224 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...