2-Chloro-4-ethynylbenzoic acid 97%

Supplier: Ambeed
Warning

A284623-100MG A284623-1G A284623-250MG
77277-024EA 23 USD
77277-024 77277-026 77277-028
2-Chloro-4-ethynylbenzoic acid 97%
2-Chloro-4-ethynylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₅ClO₂
CAS Number: 2168559-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR