4,4'-(Ethyne-1,2-diyl)dianiline 95%

Supplier: Ambeed
Danger

A284571-100MG A284571-1G A284571-250MG
77289-724EA 47.38 USD
77289-724 77289-726 77289-728
4,4'-(Ethyne-1,2-diyl)dianiline 95%
4,4'-(Ethyne-1,2-diyl)dianiline

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂
MW: 208.25 g/mol
MDL Number: MFCD00458961
CAS Number: 6052-15-9
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR