N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
Dimethylanilinium tetrakis (pentafluorophenyl)borate, N,N-Dimethylanilinium tetra(pentafluorophenyl)borate, N,N-Dimethylbenzenaminium tetrakis(perfluorophenyl)borate
A284357-100MG
A284357-1G
A284357-250MG
A284357-5G
A284357-25G
77238-944EA
6
USD
77238-944
77238-946
77238-948
77238-950
77746-520
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate 97%
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate
Precaución: for Research Use Only
Formula:
C₃₂H₁₂BF₂₀N MW: 801.23 g/mol |
MDL Number:
MFCD12913898 CAS Number: 118612-00-3 UN: 1759 ADR: 8 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...