Isopropoxybenzene 97%

Supplier: Ambeed
Warning

A284305-100G A284305-1G A284305-25G A284305-5G
77238-936EA 124.63 USD
77238-936 77238-938 77238-940 77238-942
Isopropoxybenzene 97%
Isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₂O
MW: 136.19 g/mol
MDL Number: MFCD00048288
CAS Number: 2741-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR