4-(Pyrrolidin-1-yl)benzonitrile 98%

Supplier: Ambeed
Warning

A284225-1G A284225-25G A284225-5G
77289-708EA 18 USD
77289-708 77289-710 77289-712
4-(Pyrrolidin-1-yl)benzonitrile 98%
4-(Pyrrolidin-1-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂
MDL Number: MFCD07368512
CAS Number: 10282-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR