DASA-58 98%

Supplier: Ambeed
Warning

A284203-10MG A284203-1MG A284203-25MG A284203-5MG A284203-50MG
77238-928EA 49 USD
77238-928 77238-930 77238-932 77238-934 77746-519
DASA-58 98%
DASA-58

Precaución: for Research Use Only

Formula: C₁₉H₂₃N₃O₆S
MDL Number: MFCD29472280
CAS Number: 1203494-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR