Lenvatinib mesylate 99%

Supplier: Ambeed
Warning

A283982-100MG A283982-1G A283982-250MG A283982-50MG A283982-5G A283982-25G
77238-918EA 10 USD
77238-918 77238-920 77238-922 77238-924 77238-926 77746-518
Lenvatinib mesylate 99%
Lenvatinib mesylate

Precaución: for Research Use Only

Formula: C₂₂H₂₃ClN₄O₇S
MDL Number: MFCD18633219
CAS Number: 857890-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR